missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG4 Fc Secondary Antibody (5C7cc), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-7081V
This item is not returnable.
View return policy
Description
IgG4 Fc Monoclonal antibody specifically detects IgG4 Fc in Human samples. It is validated for ELISA
Specifications
| IgG4 Fc | |
| Monoclonal | |
| DyLight 405 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 5C7cc | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with IgG4 from human serum. | |
| 0.2 mL | |
| Immunology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction