Learn More
Invitrogen™ Mouse anti-Human IgG2 Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ 053500
Product Code. 10727043
  • £252.00 / 500µg
For Research Use Only. All usage must comply with product instructions.
Description
Description
Mouse anti-Human IgG2 Secondary Antibody for ELISA, IHC (P), IP, Western Blot
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgG2
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide; pH 7.4
Human IgG2.
500 μg
Secondary
-20°C
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
HP6002
Unconjugated
Mouse
Purified
RUO
Human
Secondary Antibody
IgG1