missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG2 Fc Secondary Antibody (52G1), DyLight 488, Novus Biologicals™

Product Code. 18700772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700772 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700772 Supplier Novus Biologicals Supplier No. NB1107082G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2 Fc Monoclonal antibody specifically detects IgG2 Fc in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2 Fc
Applications ELISA
Classification Monoclonal
Clone 52G1
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with human IgG2 from myeloma patient serum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.