missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MH1715
This item is not returnable.
View return policy
Description
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgG1 Fc | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol, 4-5mg/mL BSA and 0.1% Proclin 300 | |
| Human IgG1 | |
| 500 μL | |
| Secondary | |
| 4° C | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Western Blot | |
| HP6070 | |
| HRP | |
| Mouse | |
| Purified | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction