missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2046IR
This item is not returnable.
View return policy
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion
Specifications
| IgG1 Fc | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum. | |
| 0.2 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion | |
| 2C11cc | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunodiffusion | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction