missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human IgG (Heavy Chain) Secondary Antibody
Description
Application notes: Indirect ELISA.
Antibodies are comprised of heavy and light chains. A state of disease can occur if the heavy chain of IgG is overproduced. Diseases associated with gamma heavy chain over production mainly affects older adults and has similarity to malignant lymphoma. On the other hand, some individuals that are affected by gamma heavy chain over production may be asymptomatic.
Specifications
Specifications
| Antigen | Human IgG (Heavy Chain) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 54-76 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Host Species | Mouse |
| Immunogen | Native protein. |
| Purification Method | Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?