missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals NBP1-73609
262.65 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Dot Blot.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Rhodamine | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Dot Blot | |
| LYOPH | |
| Immunocytochemistry/Immunofluorescence 1:500-1:2500, Dot Blot | |
| Human IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This product was prepared from monospecific polyclonal ascites by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum, Human IgG and Human Serum. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction