missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG Fd Secondary Antibody, PE

Mouse Monoclonal Secondary Antibody

Brand:  Invitrogen™ SA510191

Product Code. 15396331

  • £244.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SA5-10191 targets Human IgG, Fd in FACS applications and shows reactivity with Human samples. The SA5-10191 immunogen is human IgG Fd.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human IgG Fd
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
Human IgG Fd.
50 μg
Secondary
4°C, do not freeze
Liquid
Flow Cytometry
HP6045
PE
Mouse
Protein G
RUO
Human
Secondary Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.