Learn More
Mouse anti-Human IgG Fd Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA510190
226.10 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SA5-10190 targets Human IgG, Fd in ELISA, FACS, and WB applications and shows reactivity with Human samples. The SA5-10190 immunogen is human IgG Fd.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgG Fd | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol, 4-5mg/mL BSA and no preservative | |
Human IgG Fd | |
100 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2a |
ELISA, Flow Cytometry, Western Blot | |
HP6045 | |
HRP | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |