missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Fc Secondary Antibody (EM-07), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45061
This item is not returnable.
View return policy
Description
IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| IgG Fc | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1 to 4 μg/mL, ELISA 1:2000, Immunocytochemistry/Immunofluorescence | |
| Fusion protein of human IgG Fc fragment. | |
| 0.1 mg | |
| Secondary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| EM-07 | |
| FITC | |
| Mouse | |
| Size Exclusion Chromatography | |
| RUO | |
| This secondary antibody reacts with Fc part of human IgG heavy chain and with isolated Fc fragments. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction