missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG Fc Secondary Antibody, Biotin

Product Code. 15993134
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
0.2mg
Product Code. Quantity unitSize
15993134 500 μL 0.2mg
1 options
This item is not returnable. View return policy

Product Code. 15993134

Brand: Invitrogen™ 054240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Secondary Antibody

Reconstitute with 0.5 mL of distilled water.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG Fc
Applications ELISA
Classification Monoclonal
Clone HP6017
Conjugate Biotin
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.4
Host Species Mouse
Immunogen Gamma Immunoglobin.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.