missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-467B
This item is not returnable.
View return policy
Description
IgG F(ab) Monoclonal antibody specifically detects IgG F(ab) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| IgG F(ab) | |
| Monoclonal | |
| Biotin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 4A11 | |
| PBS | |
| Purified human IgG. | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction