Learn More
Mouse anti-Human IgE Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA182854
Code : LD
Additional Details : Weight : 0.01000kg
Description
Mouse anti Human IgE antibody, clone E411 (5H2) recognizes human IgE heavy chain, a 428 amino acid protein bearing four Ig-like domains. Mouse anti Human IgE antibody, clone E411 binds to an epitope expressed on Cε3 domain.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgE | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide; pH 7.2 | |
Immunoglobulin E from human serum. | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2a |
ELISA, Flow Cytometry, Western Blot | |
E411 (5H2) | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |