Learn More
Invitrogen™ Mouse anti-Human IgE Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ SA119259
Product Code. 11505030
  • £278.00 / 100µg
Description
This antibody recognizes an epitope different from the ones recognized by 4G7 and 4H10 antibodies to IgE.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgE
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
Purified human IgE.
100 μg
Secondary
4°C, do not freeze
Liquid
ELISA, Flow Cytometry
BE5
Unconjugated
Mouse
Protein A
RUO
Human
Secondary Antibody
IgG1