missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgE, Clone: G7-26, biotin, Secondary Antibody, BD
Brand: BD Biosciences 555858
Additional Details : Weight : 0.01000kg
Description
The G7-26 antibody reacts with human immunoglobulin E. It does not react with other heavy chain isotypes.
Host Species: Mouse
Clone: G7-26
Isotype: IgG2a, κ
Species Reactivity: Human
ELISA
Specifications
Mouse anti-Human IgE | |
G7-26 | |
Biotin | |
0.5 mg | |
Immunology | |
Human |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG2a κ |