missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
£414.00 - £828.00
Specifications
| Antigen | Human Ig Mix |
|---|---|
| Clone | HIG10-101.1.19 |
| Concentration | 1.0 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11515000
|
Invitrogen™
SA110648 |
500 μg |
£414.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17810446
|
Invitrogen™
SA5-10348 |
1 mg |
£828.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17820306
|
Invitrogen™
SA5-10349 |
100 μg |
£828.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SA1-10648 detects human Immunoglobulin. SA1-10648 has been successfully used in ELISA applications. The SA1-10648 immunogen is purified human immunoglobulin.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Human Ig Mix | |
| 1.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Secondary | |
| Protein A |
| HIG10-101.1.19 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| Purified human immunoglobulin. | |
| Secondary Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title