Learn More
CD141 Mouse anti-Human, Brilliant Violet 786, Clone: 1A4, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741006
Description
Reacts with a 75 kDa, a single chain, type I membrane glycoprotein referred to as thrombomodulin and fetomodulin. CD141 is expressed on endothelial cells, smooth muscle cells, epithelial cells, synovial lining cells, and keratinocytes. It is also found on megakaryocytes, peripheral blood moncytes, and neutrophils, but not on lymphocytes. Reports indicate that CD 141 plays an important role in Protein C activation and the initiation of the Protein C anticoagulant pathway. Thrombin loses its procagulant function when it binds to CD141, and the CD141/thrombin complex is able to activate Protein C.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
| CD141 | |
| Monoclonal | |
| 0.2mg/mL | |
| Thrombomodulin | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1A4 | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only