Learn More
HSP90AB1, Mouse, Clone: MBH90B, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of HSP90AB1.
Brand: Abnova MAB6458.100ug
Description
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1 (MIM 140571), an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM
Sequence: EEVHHGEEEVECSpecifications
| HSP90AB1 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (1 ug/mL) Western Blot (1 ug/mL) Immunoprecipitation (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| HSP90AB1 | |
| HSP90AB1 | |
| A synthetic peptide corresponding to amino acids at N-terminus of human HSP90AB1. | |
| RUO | |
| 3326 | |
| Bovine, Human, Mouse | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| MBH90B | |
| Mouse monoclonal antibody raised against synthetic peptide of HSP90AB1. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| D6S182/FLJ26984/HSP90-BETA/HSP90B/HSPC2/HSPCB | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes the EEVHHG epitope within the N-terminal part of Hsp90b, an ubiquitously expressed protein with calculated MW of 83.3 KDa, however, migrating as a 90 KDa band under reducing SDS-PAGE conditions. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.