Learn More
HR, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HR.
Brand: Abnova H00055806-A01.50uL
Description
This gene encodes a protein that is involved in hair growth. This protein functions as a transcriptional corepressor of multiple nuclear receptors, including thyroid hormone receptor, the retinoic acid receptor-related orphan receptors and the vitamin D receptors, and it interacts with histone deacetylases. The translation of this protein is modulated by multiple regulatory ORFs that exist upstream of the primary ORF. Mutations in one of these upstream ORFs, U2HR, cause Marie Unna hereditary hypotrichosis (MUHH), an autosomal dominant form of genetic hair loss. Mutations in this gene also cause autosomal recessive congenital alopecia and atrichia with papular lesions, other diseases resulting in hair loss. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: LDAGLRRRLREEWGVSCWTLLQAPGEAVLVPAGAPHQVQGLVSTVSVTQHFLSPETSALSAQLCHQGPSLPPDCHLLYAQMDWAVFQAVKVAVGTLQEAKSpecifications
| HR | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant HR. | |
| HR | |
| ALUNC/AU/HSA277165 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005144 | |
| HR | |
| HR (NP_005135, 1090 a.a. ∽ 1189 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 55806 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only