missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-HPRT, Clone: 2G8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74045-100UL
This item is not returnable.
View return policy
Description
HPRT Monoclonal specifically detects HPRT in Human samples. It is validated for Western Blot, ELISA.
Specifications
| HPRT | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:5000, ELISA 1:100-1:2000 | |
| NP_000185 | |
| HPRT1 | |
| Recombinant human HPRT (1-218aa) purified from E. coli | |
| 100 μL | |
| Lipid and Metabolism | |
| 3251 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2G8 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human HPRT mAb, clone 2G8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HPRT protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction