Learn More
HNF4A, Mouse, Clone: 4E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HNF4A.
Brand: Abnova H00003172-M04A.200uL
Description
The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal dominant noninsulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Sequence: NDRQYDSRGRFGELLLLLPTLQSITWQMIEQIQFIKLFGMAKIDNLLQEMLLGGSPSDAPHAHHPLHPHLMQEHMGTNVIVANTMPTHLSNGQMCEWPRPSpecifications
| HNF4A | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_000457.4 | |
| HNF4A | |
| HNF4A (NP_000448.3,324 a.a. ∽ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3172 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4E2 | |
| Mouse monoclonal antibody raised against a partial recombinant HNF4A. | |
| HNF4A | |
| FLJ39654/HNF4/HNF4a7/HNF4a8/HNF4a9/MODY/MODY1/NR2A1/NR2A21/TCF/TCF14 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.