missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR/HLA-DP, Mouse, Clone: MEM-136, Abnova™
Mouse monoclonal antibody raised against native HLA-DR/HLA-DP.
Brand: Abnova MAB0917.100ug
This item is not returnable.
View return policy
Specifications
| HLA-DR/HLA-DP | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| DPB1/HLA-DP1B | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes common epitope on beta-chain of human HLA-DR and HLA-DP. It reacts with alpha/beta dimer as well as with dissociated beta-subunit. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| MEM-136 | |
| Mouse monoclonal antibody raised against native HLA-DR/HLA-DP. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| HLA-DPB1 | |
| Native purified HLA-DR/HLA-DP from PHA-activated peripheral blood leucocytes. | |
| RUO | |
| 3115/3123/3125/3126/3127 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction