missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-A/HLA-B/HLA-C, Mouse, Clone: LY5.1, Abnova™
Mouse monoclonal antibody raised against HLA-A/HLA-B/HLA-C.
Brand: Abnova MAB6383.500ug
This item is not returnable.
View return policy
Specifications
| HLA-A/HLA-B/HLA-C | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| HLAA | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3105/3106/3107 | |
| Store at -20°C. After reconstitution with deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunohistochemistry | |
| LY5.1 | |
| Mouse monoclonal antibody raised against HLA-A/HLA-B/HLA-C. | |
| In 10mM PBS, pH 7.2 | |
| HLA-A | |
| Purified human HLA-A/HLA-B/HLA-C complex. | |
| 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction