missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 p24 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against full-length recombinant HIV-1 p24.
Brand: Abnova MAB11187.100ug
This item is not returnable.
View return policy
Specifications
| HIV-1 p24 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (0.5-0.2 ug/mL) ELISA (0.5-0.2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Peptide affinity purification | |
| RUO | |
| By Western blot, anti-HIV-1 p24 antibody detects a ∼24 kDa, a ∼41 kDa, and a ∼55 kDa protein, corresponding to HIV-1 p24 and to its precursors p41 and p55, respectively, in HIV-1 samples. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against full length recombinant HIV-1 p24. | |
| In PBS (0.02% sodium azide) | |
| Recombinant protein corresponding to human immunodeficiency virus type 1 p24. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction