missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1R, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against HIP1R.
Brand: Abnova MAB6239.100uL
This item is not returnable.
View return policy
Specifications
| HIP1R | |
| Monoclonal | |
| Mouse monoclonal antibody raised against HIP1R. | |
| In ascites (0.09% sodium azide) | |
| FLJ14000/FLJ27022/HIP12/HIP3/ILWEQ/KIAA0655/MGC47513 | |
| Mouse | |
| 100 μL | |
| Primary | |
| This is specific to HIP1r. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| HIP1R | |
| HIP1R | |
| Human HIP1r-TALIN Homology Region (57 kDa). | |
| RUO | |
| 9026 | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction