Learn More
HERC5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HERC5.
Brand: Abnova H00051191-A01.50uL
Description
This gene is a member of the HERC family of ubiquitin ligases and encodes a protein with a HECT domain and five RCC1 repeats. Pro-inflammatory cytokines upregulate expression of this gene in endothelial cells. The protein localizes to the cytoplasm and perinuclear region and functions as an interferon-induced E3 protein ligase that mediates ISGylation of protein targets. The gene lies in a cluster of HERC family genes on chromosome 4. [provided by RefSeq
Sequence: YDWKTFEKNARYEPGYNSSHPTIVMFWKAFHKLTLEEKKKFLVFLTGTDRLQMKDLNNMKITFCCPESWNERDPIRALTCFSVLFLPKYSTMETVEEALQEAINNNRGFGSpecifications
| HERC5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant HERC5. | |
| HERC5 | |
| CEB1/CEBP1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016323 | |
| HERC5 | |
| HERC5 (NP_057407, 915 a.a. ∽ 1024 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 51191 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only