missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B presurface antigen, Mouse, Clone: S26, Abnova™
Mouse monoclonal antibody raised against hepatitis B Presurface antigen.
Brand: Abnova MAB4776.100ug
Additional Details : Weight : 0.02000kg
Specifications
Hepatitis B presurface antigen | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Virus | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
S26 | |
Mouse monoclonal antibody raised against hepatitis B Presurface antigen. | |
In PBS, pH 7.4 | |
Hepatitis B presurface antigen from human sera pool. | |
RUO | |
HBsAg pre-S2 antigen (a.a. 132-137) (adw nomenclature). Reference start codon was Pre-S1. Epitope recognition confirmed by competing with peptides. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |