missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hcn4, Mouse, Clone: S114-10, Abnova™
Mouse monoclonal antibody raised against partial recombinant Hcn4.
Brand: Abnova MAB6667.100ug
This item is not returnable.
View return policy
Specifications
| Hcn4 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1.0-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Hcn4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects ∼130KDa. No cross-reactivity against other HCNs. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| S114-10 | |
| Mouse monoclonal antibody raised against partial recombinant Hcn4. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Hcn4 | |
| Recombinant fusion protein corresponding to C-terminus amino acids 1019-1198 of rat Hcn4. | |
| RUO | |
| 59266 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction