Learn More
HCK, Mouse, Clone: 3D12E10, Abnova™
Mouse monoclonal antibody raised against partial recombinant HCK.
Brand: Abnova MAB10679.100uL
Description
The protein encoded by this gene is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization. [provided by RefSeq
Specifications
| HCK | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| HCK | |
| HCK | |
| Recombinant protein corresponding to human HCK. | |
| RUO | |
| 3055 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 3D12E10 | |
| Mouse monoclonal antibody raised against partial recombinant HCK. | |
| In ascites (0.03% sodium azide) | |
| JTK9 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.