Learn More
HBQ1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HBQ1.
Brand: Abnova H00003049-A01.50uL
Description
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5' - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3'. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue. [provided by RefSeq]
Sequence: MALSAEDRALVRALWKKLGSNVGVYTTEALERTFLAFPATKTYFSHLDLSPGSSQVRAHGQKVADALSLAVERLDDLPSpecifications
| HBQ1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant HBQ1. | |
| HBQ1 | |
| HBQ1 | |
| HBQ1 (NP_005322, 1 a.a. ∽ 78 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 3049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005331 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only