Learn More
HAX1, Mouse, Clone: 9G3D11, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of HAX1.
Brand: Abnova MAB7876.100ug
Description
The protein encoded by this gene is known to associate with hematopoietic cell-specific Lyn substrate 1, a substrate of Src family tyrosine kinases. It also interacts with the product of the polycystic kidney disease 2 gene, mutations in which are associated with autosomal-dominant polycystic kidney disease, and with the F-actin-binding protein, cortactin. It was earlier thought that this gene product is mainly localized in the mitochondria, however, recent studies indicate it to be localized in the cell body. Mutations in this gene result in autosomal recessive severe congenital neutropenia, also known as Kostmann disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| HAX1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| HAX1 | |
| HAX1 | |
| A synthetic peptide corresponding to N-terminus 15 amino acids of human HAX1. | |
| RUO | |
| 10456 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 9G3D11 | |
| Mouse monoclonal antibody raised against synthetic peptide of HAX1. | |
| In PBS (0.02% sodium azide) | |
| FLJ17042/FLJ18492/FLJ93803/HCLSBP1/HS1BP1/SCN3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.