Learn More
GTF3A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GTF3A.
Brand: Abnova H00002971-A01.50uL
Description
The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein. [provided by RefSeq
Sequence: NQQKQYICSFEDCKKTFKKHQQLKIHQCQNTNEPLFKCTQEGCGKHFASPSKLKRHAKAHEGYVCQKGCSFVAKTWTELLKHVRETHKEESpecifications
| GTF3A | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GTF3A. | |
| GTF3A | |
| AP2/TFIIIA | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002097 | |
| GTF3A | |
| GTF3A (NP_002088, 185 a.a. to 274 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 2971 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only