Learn More
GSTP1, Mouse, Clone: Usal-hGST-Pi-McAb-1, Abnova™
Mouse monoclonal antibody raised against recombinant GSTP1.
Brand: Abnova MAB3452.100ug
Description
Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases. [provided by RefSeq
Specifications
| GSTP1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user. | |
| GSTP1 | |
| GSTP1 | |
| Recombinant protein corresponding to human GSTP1. | |
| RUO | |
| 2950 | |
| Human, Leishmania species, Rat | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| Usal-hGST-Pi-McAb-1 | |
| Mouse monoclonal antibody raised against recombinant GSTP1. | |
| In buffer containing 100nM sodium azide | |
| DFN7/FAEES3/GST3/PI | |
| Mouse | |
| 100 μg | |
| Primary | |
| Recognizes human, rat and leishmania GST-Pi. Does not cross-react with human or rat alpha GST. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.