Learn More
GRK1, Mouse, Clone: 4E9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GRK1.
Brand: Abnova H00006011-M02A.200uL
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates rhodopsin and initiates its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). [provided by RefSeq
Sequence: RDSLSLEFESVCLEQPIGKKLFQQFLQSAEKHLPALELWKDIEDYDTADNDLQPQKAQTILAQYLDPQAKLFCSFLDEGIVAKFKEGPVEIQDGLFQPLLSpecifications
| GRK1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_002929 | |
| GRK1 | |
| GRK1 (NP_002920.1,51 a.a. ∽ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6011 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4E9 | |
| Mouse monoclonal antibody raised against a partial recombinant GRK1. | |
| GRK1 | |
| GPRK1/RHOK/RK | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.