missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grin1, Mouse, R-PE, Clone: S308-48, Abnova™
Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
Brand: Abnova MAB11726.100ug
This item is not returnable.
View return policy
Specifications
| Grin1 | |
| Monoclonal | |
| R-PE | |
| Immunohistochemistry (0.1-1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Grin1 | |
| Grin1 | |
| Recombinant fusion protein corresponding to amino acids 42-361 at extracellular N-terminus of rat Grin1. | |
| 100 μg | |
| Primary | |
| Rat | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| S308-48 | |
| Mouse monoclonal antibody raised against partial synthetic protein of Grin1. | |
| In PBS pH 7.4 (50% glycerol and 0.09% sodium azide) | |
| NMDAR1/NR1 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 24408 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction