Learn More
GRAP2, Mouse, Clone: UW40, Abnova™
Mouse monoclonal antibody raised against recombinant GRAP2.
Brand: Abnova MAB10180.100ug
Description
This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist. [provided by RefSeq
Specifications
| GRAP2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1 ug/ml) The optimal working dilution should be determined by the end user. | |
| GRAP2 | |
| GRAP2 | |
| Recombinant GST fusion protein corresponding to human GRAP2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| UW40 | |
| Mouse monoclonal antibody raised against recombinant GRAP2. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| GADS/GRAP-2/GRB2L/GRBLG/GRID/GRPL/GrbX/Grf40/Mona/P38 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 9402 | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.