missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Goat IgG (H+L) Secondary Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals NBP1-73796
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Mouse | |
| Delipidation and Defibrination | |
| RUO | |
| Secondary | |
| Reconstitute with 2.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Goat IgG whole molecule | |
| 2 mL | |
| Adaptive Immunity, Immunology | |
| This product was prepared from monospecific ascites by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against Goat IgG and Goat Serum. | |
| Goat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction