Learn More
GNMT, Mouse, Clone: 2B10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GNMT.
Brand: Abnova H00027232-M01A.200uL
Description
The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine (along with glycine) to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency (hypermethioninemia). [provided by RefSeq
Sequence: MVDSVYRTRSLGVAAEGLPDQYADGEAARVWQLYIGDTRSRTAEYKAWLLGLLRQHGCQRVLDVACGTGVDSIMLVEEGFSVTSVDASDKMLKYALKERWSpecifications
| GNMT | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_018960 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA | |
| 2B10 | |
| Mouse monoclonal antibody raised against a partial recombinant GNMT. | |
| GNMT | |
| GNMT | |
| GNMT (NP_061833.1,1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 27232 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.