Learn More
GNG3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant GNG3.
Brand: Abnova H00002785-A01.50uL
Description
G proteins are heterotrimers of alpha, beta, and gamma subunits. Gamma subunits, such as GNG3, contribute to the specificity of the hundreds of receptor signaling pathways involving G proteins (Schwindinger et al., 2004 [PubMed 15314181]).
Sequence: MKGETPVNSTMSIGQARKMVEQLKIEASLCRIKVSKAAADLVTYCDAHACEDPLITPVPTSENPFREKKFFCALLSpecifications
| GNG3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant GNG3. | |
| GNG3 | |
| GNG3 | |
| GNG3 (AAH15563, 1 a.a. to 75 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 2785 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC015563 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only