Learn More
GLTP, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GLTP.
Brand: Abnova H00051228-A01.50uL
Description
The protein encoded by this gene is similar to bovine and porcine proteins which accelerate transfer of certain glycosphingolipids and glyceroglycolipids between membranes. It is thought to be a cytoplasmic protein. [provided by RefSeq
Sequence: SICDGERDENHPNLIRVNATKAYEMALKKYHGWIVQKIFQAALYAAPYKSDFLKALSKGQNVTEEECLEKIRLFLVNYTATIDVIYEMYTQMNAELNYKVSpecifications
| GLTP | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GLTP. | |
| GLTP | |
| GLTP | |
| GLTP (NP_057517, 110 a.a. to 209 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 51228 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016433 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only