Learn More
GHRL, Mouse, Clone: 2E2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GHRL.
Brand: Abnova H00051738-M05A.200uL
Description
This gene encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Obestatin was initially reported to be an endogenous ligand for the orphan G protein-coupled receptor GPR39 and was involved in satiety and decreased food intake; however, these findings are controversial. Recent reports show that obestatin is involved in inhibiting thirst and anxiety, improving memory, regulating sleep, affecting cell proliferation, and increasing the secretion of pancreatic juice enzymes. Alternative promoters and alternative splicing result in multiple transcript variants, some of which encode different protein isoforms and some of which do not encode a protein but may regulate the ghrelin-obestatin preproprotein expression. In addition, antisense transcripts for this gene have been identified and may also function in regulation of the ghrelin-obestatin preproprotein expression. [provided by RefSeq
Sequence: MPSPGTVCSLLLLGMLWLDLAMAGSSFLSPEHQRVQQRKESKKPPAKLQPRALAGWLRPEDGGQAEGAEDEMEVRFNAPFDVGIKLSGVQYQQHSQALGKFLQDILWEEAKEAPADKSpecifications
| GHRL | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC025791 | |
| GHRL | |
| GHRL (AAH25791.1,1 a.a. ∽ 117 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51738 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2E2 | |
| Mouse monoclonal antibody raised against a full-length recombinant GHRL. | |
| GHRL | |
| MTLRP/obestatin | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.