missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP, Mouse, Clone: GF-01, Abnova™
Mouse monoclonal antibody raised against native GFAP.
Brand: Abnova MAB3612.100ug
This item is not returnable.
View return policy
Specifications
| GFAP | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| GFAP | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with GFAP, the principal marker of astroglial cells in the central nervous system, which is specifically expressed in satellite cells in peripheral ganglia and in non myelinating Schwann cells in peripheral nerves. The GFAP protein runs on gels at ∼55 KDa protein, usually associated with lower Mw bands which are thought to be proteolytic fragments and alternate transcripts from the single gene. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| GF-01 | |
| Mouse monoclonal antibody raised against native GFAP. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| GFAP | |
| Native purified porcine GFAP. | |
| RUO | |
| 396562 | |
| Feline, Human, Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction