missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against of GAPDH.
Brand: Abnova MAB11147.100ug
This item is not returnable.
View return policy
Specifications
| GAPDH | |
| Monoclonal | |
| Mouse monoclonal antibody raised against of GAPDH. | |
| In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) | |
| Purified pig GAPDH. | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-4000) Immunofluorescence (1:100-400) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein G purification | |
| RUO | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction