Learn More
GAD1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant GAD1.
Brand: Abnova H00002571-A01.50uL
Description
This gene encodes one of several forms of glutamic acid decarboxylase, identified as a major autoantigen in insulin-dependent diabetes. The enzyme encoded is responsible for catalyzing the production of gamma-aminobutyric acid from L-glutamic acid. A pathogenic role for this enzyme has been identified in the human pancreas since it has been identified as an autoantigen and an autoreactive T cell target in insulin-dependent diabetes. This gene may also play a role in the stiff man syndrome. Deficiency in this enzyme has been shown to lead to pyridoxine dependency with seizures. Alternative splicing of this gene results in two products, the predominant 67kDa form and a less-frequent 25kDa form. [provided by RefSeq
Sequence: MASSTPSSSATSSNAGADPNTTNLRPTTYDTWCGVAHGCTRKLGLKICGFLQRTNSLEEKSRLVSAFKERQSSKNLLSCENSDRDARFRRTETDFSNLFARDLLPAKNGEEQTVQFLLEVVDILLNYVRKTFDRSTKVLDFHHPHQLLEGMEGFNLELSDHPESLEQILVDCRDTLKYGVRTGHPRFFNQLSTGLDIIGLAGEWLTSTANTNMFTYEIAPVFVLMEQITLKKMREIVGWSSKDGDGIFSPGGAISNMYSIMAARYKYFPEVKTKGMAAVPKLVLFTSEQSHYSIKKAGAALGFGTDNVILIKCNERGKIIPADFEAKILEAKQKGYVPFYVNATAGTTVYGAFDPIQEIADICEKYNLWLHVDAAWGGGLLMSRKHRHKLNGIERANSVTWNPHKMMGVLLQCSAILVKEKGILQGCNQMCAGYLFQPDKQYDVSYDTGDKAIQCGRHVDIFKFWLMWKAKGTVGFENQINKCLELAEYLYAKIKNREEFEMVFNGEPEHTNVCFWYIPQSLRGVPDSPQRREKLHKVAPKIKALMMESGTTMVGYQPQGDKANFFRMVISNPAATQSDIDFLIEEIERLGQDLSpecifications
| GAD1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant GAD1. | |
| GAD1 | |
| FLJ45882/GAD/SCP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC026349 | |
| GAD1 | |
| GAD1 (AAH26349, 1 a.a. to 594 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 2571 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only