missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRK, Mouse, Clone: 4E1, Abnova™
Mouse monoclonal antibody raised against partial recombinant FRK.
Brand: Abnova MAB10656.100uL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth. [provided by RefSeq
Specifications
| FRK | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| FRK | |
| FRK | |
| Recombinant protein corresponding to human FRK. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 4E1 | |
| Mouse monoclonal antibody raised against partial recombinant FRK. | |
| In PBS (50% glycerol, 0.03% sodium azide) | |
| GTK/PTK5/RAK | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2444 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction