missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ35767, Mouse, Clone: 2F3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FLJ35767.
Brand: Abnova H00400629-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MCPPVSMRYEEEGMSYLYASWMYQLQHGDQLSICFTCFKAAFLDFKDLLESEDWEEDNWDPELMEHTEAESEQEGSSGMELSWGQSPGQPVQGGSEAWGPGTLAAAPEGLEDAGLDPHFVPTELWPQEAVPLGLGLEDADWTQGLPWRFEELLTCSHWPSFFPSSpecifications
| FLJ35767 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_207459.1 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 400629 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| ELISA, Western Blot | |
| 2F3 | |
| Mouse monoclonal antibody raised against a full-length recombinant FLJ35767. | |
| FLJ35767 | |
| FLJ35767 | |
| FLJ35767 (NP_997342.1,1 a.a. ∽ 164 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction