Learn More
FGFR4, Mouse, Clone: 7H1, Abnova™
Mouse monoclonal antibody raised against recombinant FGFR4.
Brand: Abnova MAB10427.100uL
Description
The protein encoded by this gene is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. The genomic organization of this gene, compared to members 1-3, encompasses 18 exons rather than 19 or 20. Although alternative splicing has been observed, there is no evidence that the C-terminal half of the IgIII domain of this protein varies between three alternate forms, as indicated for members 1-3. This particular family member preferentially binds acidic fibroblast growth factor and, although its specific function is unknown, it is overexpressed in gynecological tumor samples, suggesting a role in breast and ovarian tumorigenesis. [provided by RefSeq
Specifications
| FGFR4 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| FGFR4 | |
| FGFR4 | |
| Recombinant protein corresponding to human FGFR4. | |
| RUO | |
| 2264 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Western Blot | |
| 7H1 | |
| Mouse monoclonal antibody raised against recombinant FGFR4. | |
| In ascitic (0.03% sodium azide) | |
| CD334/JTK2/MGC20292/TKF | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.