Learn More
FGF21, Mouse, Clone: 3E20, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FGF21.
Brand: Abnova H00026291-M06A.200uL
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined. [provided by RefSeq
Sequence: PIPDSSPLLQFGGQVRQRYLYTDDAQQTEAHLEIREDGTVGGAADQSPESLLQLKALKPGVIQILGVKTSRFLCQRPDGALYGSLHFDPEACSFRELLLEDGYNVYQSEAHGLPLHLPGNKSPHRDPAPRGPARFLPLPGLPPAPPEPPGILAPQPPDVGSSDPLSMVGPSQGRSPSYASSpecifications
| FGF21 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC018404 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA, Western Blot | |
| 3E20 | |
| Mouse monoclonal antibody raised against a full-length recombinant FGF21. | |
| FGF21 | |
| FGF21 | |
| FGF21 (AAH18404,30 a.a. ∽ 209 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 26291 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.