missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Feline Ig Mix Secondary Antibody

Mouse Monoclonal Secondary Antibody

£383.00 - £828.00

Specifications

Antigen Feline Ig Mix
Clone FIG11-207.2
Concentration 0.6 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11574990
View Documents
Invitrogen™
SA110643
500 μg
£383.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17880386
View Documents
Invitrogen™
SA5-10354
100 μg
£828.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17830306
View Documents
Invitrogen™
SA5-10355
1 mg
£828.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SA1-10643 detects feline Immunoglobulin. Does not react with bovine, equine, canine, rabbit, sheep, porcine, donkey, guinea pig, goat or human. SA1-10643 has been successfully used in ELISA applications. The SA1-10643 immunogen is purified feline immunoglobulin.

Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Feline Ig Mix
0.6 mg/mL
ELISA
Unconjugated
Mouse
RUO
PBS with no preservative; pH 7.4
Secondary
Protein A
FIG11-207.2
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG1
Feline
Purified feline immunoglobulin.
Secondary Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.