missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR3B, Mouse, Biotin, Clone: MEM-154, Abnova™
Mouse monoclonal antibody raised against native FCGR3B.
Brand: Abnova MAB4538.100ug
This item is not returnable.
View return policy
Specifications
| FCGR3B | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (1:500) The optimal working dilution should be determined by the end user. | |
| FCGR3B | |
| FCGR3B | |
| Native purified FCGR3B from human granulocytes. | |
| RUO | |
| 2215 | |
| Human | |
| Liquid |
| Flow Cytometry | |
| MEM-154 | |
| Mouse monoclonal antibody raised against native FCGR3B. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| CD16/CD16b/FCG3/FCGR3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with the epitope on CD16 antigen that residing in proximity to FG loop (probably BC or C'E loop). This antibody reacts with CD16+ granulocytes. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction